3-Chloro-2,4-dimethylbenzoyl chloride ≥95%

Supplier: AOBChem USA

Warning

79476-5G 79476-25G 79476-1G 79476-500MG
77630-674EA 1441 USD
77630-674 77660-358 77644-022 77647-996
3-Chloro-2,4-dimethylbenzoyl chloride ≥95%
3-Chloro-2,4-dimethylbenzoyl chloride

Precaución: For Research Use Only

Formula: C₉H₈Cl₂O
Storage Temperature: Ambient
MDL Number: MFCD34767985
CAS Number: 2386816-48-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR