1-Iodo-3-phenoxybenzene ≥95%

Supplier: AOBChem USA

Warning

79413-5G 79413-1G 79413-500MG 79413-25G
77633-720EA 1129 USD
77633-720 77639-996 77653-084 77660-356
1-Iodo-3-phenoxybenzene ≥95%
1-Iodo-3-phenoxybenzene

Precaución: For Research Use Only

Formula: C₁₂H₉IO
Storage Temperature: Ambient
MDL Number: MFCD18207352
CAS Number: 2974-95-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR