2-Bromo-4-fluoro-6-hydroxybenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

79387-5G 79387-1G 79387-500MG
77660-354EA 2525 USD
77660-354 77647-992 77647-994
2-Bromo-4-fluoro-6-hydroxybenzaldehyde ≥95%
2-Bromo-4-fluoro-6-hydroxybenzaldehyde

Precaución: For Research Use Only

Formula: C₇H₄BrFO₂
Storage Temperature: Ambient
MDL Number: MFCD23705539
CAS Number: 1427382-05-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR