Methyl 3-bromo-5-methoxy-2-methylbenzoate ≥95%

Supplier: AOBChem USA

Warning

79354-5G 79354-25G 79354-1G 79354-500MG
77633-718EA 1407 USD
77633-718 77656-892 77644-020 77647-990
Methyl 3-bromo-5-methoxy-2-methylbenzoate ≥95%
Methyl 3-bromo-5-methoxy-2-methylbenzoate

Precaución: For Research Use Only

Formula: C₁₀H₁₁BrO₃
Storage Temperature: Ambient
MDL Number: MFCD27944540
CAS Number: 13979-62-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR