3-Bromo-5-methoxy-2-methylbenzoic acid ≥95%

Supplier: AOBChem USA

Warning

79353-1G 79353-500MG 79353-5G
77635-310EA 861 USD
77635-310 77653-082 77656-890
3-Bromo-5-methoxy-2-methylbenzoic acid ≥95%
3-Bromo-5-methoxy-2-methylbenzoic acid

Precaución: For Research Use Only

Formula: 245.07 g/mol
MW: C₉H₉BrO₃
Storage Temperature: Ambient
MDL Number: MFCD00267662
CAS Number: 13979-61-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR