2',4',5'-Trimethylacetophenone ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 1-(2,4,5-Trimethylphenyl)ethan-1-one, 1-(2,4,5-Trimethylphenyl)ethanone

78009-5G 78009-25G
77635-308EA 59 USD
77635-308 77660-342
2',4',5'-Trimethylacetophenone ≥95%
2',4',5'-Trimethylacetophenone

Precaución: For Research Use Only

Formula: C₁₁H₁₄O
MW: 162.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00017244
CAS Number: 2040-07-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR