Methyl(2,4,5-trimethylphenyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

78007-5G 78007-1G 78007-500MG
77660-340EA 1961 USD
77660-340 77644-014 77647-980
Methyl(2,4,5-trimethylphenyl)sulfane ≥97%
Methyl(2,4,5-trimethylphenyl)sulfane

Precaución: For Research Use Only

Formula: 166.2831 g/mol
MW: C₁₀H₁₄S
Storage Temperature: Ambient
MDL Number: MFCD11617767
CAS Number: 1215002-19-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR