3-Bromo-5-chloro-2-ethoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

77921-5G 77921-25G 77921-1G 77921-500MG
77630-670EA 1441 USD
77630-670 77660-338 77644-012 77647-978
3-Bromo-5-chloro-2-ethoxybenzaldehyde ≥97%
3-Bromo-5-chloro-2-ethoxybenzaldehyde

Precaución: For Research Use Only

Formula: C₉H₈BrClO₂
Storage Temperature: Ambient
MDL Number: MFCD09721052
CAS Number: 540779-02-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR