3,6-Dichloro-2-picoline ≥97%

Supplier: AOBChem USA

Warning

77893-5G 77893-25G
77635-306EA 114 USD
77635-306 77656-888
3,6-Dichloro-2-picoline ≥97%
3,6-Dichloro-2-picoline

Precaución: For Research Use Only

Formula: C₆H₅Cl₂N
Storage Temperature: Ambient
MDL Number: MFCD00778766
CAS Number: 123280-64-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR