1-(Benzo[b]thiophen-2-yl)-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

77827-1G 77827-5G 77827-500MG
77635-304EA 1003 USD
77635-304 77660-336 77652-004
1-(Benzo[b]thiophen-2-yl)-2,2-difluoroethanone ≥95%
1-(Benzo[b]thiophen-2-yl)-2,2-difluoroethanone

Precaución: For Research Use Only

Formula: 212.216 g/mol
MW: C₁₀H₆F₂OS
Storage Temperature: Ambient
MDL Number: MFCD34767236
CAS Number: 1784765-60-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR