Methyl 5-bromo-2-isopropylbenzoate ≥97%

Supplier: AOBChem USA

Warning

77638-1G 77638-5G 77638-500MG
77639-990EA 639 USD
77639-990 77656-886 77652-002
Methyl 5-bromo-2-isopropylbenzoate ≥97%
Methyl 5-bromo-2-isopropylbenzoate

Precaución: For Research Use Only

Formula: C₁₁H₁₃BrO₂
Storage Temperature: Ambient
MDL Number: MFCD34767135
CAS Number: 2092388-13-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR