5-Bromo-2-isopropylbenzoic acid ≥95%

Supplier: AOBChem USA

Warning

77637-1G 77637-500MG 77637-5G
77639-988EA 766 USD
77639-988 77653-080 77660-330
5-Bromo-2-isopropylbenzoic acid ≥95%
5-Bromo-2-isopropylbenzoic acid

Precaución: For Research Use Only

Formula: 243.097 g/mol
MW: C₁₀H₁₁BrO₂
Storage Temperature: Ambient
MDL Number: MFCD16619422
CAS Number: 883995-21-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR