Methyl 2,6-bis(trifluoromethyl)benzoate ≥97%

Supplier: AOBChem USA

Warning

77372-5G 77372-25G 77372-1G 77372-500MG
77633-712EA 1330 USD
77633-712 77656-884 77644-010 77647-974
Methyl 2,6-bis(trifluoromethyl)benzoate ≥97%
Methyl 2,6-bis(trifluoromethyl)benzoate

Precaución: For Research Use Only

Formula: C₁₀H₆F₆O₂
Storage Temperature: Ambient
CAS Number: 34060-79-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR