5-Chloro-2-(trifluoromethoxy)benzaldehyde ≥95%

Supplier: AOBChem USA

Warning

76807-5G 76807-25G
77635-302EA 117 USD
77635-302 77660-328
5-Chloro-2-(trifluoromethoxy)benzaldehyde ≥95%
5-Chloro-2-(trifluoromethoxy)benzaldehyde

Precaución: For Research Use Only

Formula: C₈H₄ClF₃O₂
MW: 224.57 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11519353
CAS Number: 1092461-15-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR