1-Bromo-4-isobutoxy-2,5-dimethylbenzene ≥97%

Supplier: AOBChem USA

Warning

76220-5G 76220-25G 76220-1G 76220-500MG
77633-708EA 1034 USD
77633-708 77660-326 77644-004 77647-966
1-Bromo-4-isobutoxy-2,5-dimethylbenzene ≥97%
1-Bromo-4-isobutoxy-2,5-dimethylbenzene

Precaución: For Research Use Only

Formula: C₁₂H₁₇BrO
Storage Temperature: Ambient
MDL Number: MFCD34766352
CAS Number: 2901099-16-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR