1-Bromo-4-isopropoxy-2,5-dimethylbenzene ≥97%

Supplier: AOBChem USA

Warning

76219-5G 76219-500MG 76219-25G 76219-1G
77635-300EA 556 USD
77635-300 77653-078 77656-878 77644-002
1-Bromo-4-isopropoxy-2,5-dimethylbenzene ≥97%
1-Bromo-4-isopropoxy-2,5-dimethylbenzene

Precaución: For Research Use Only

Formula: C₁₁H₁₅BrO
Storage Temperature: Ambient
MDL Number: MFCD34766351
CAS Number: 1644244-99-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR