5-Ethoxy-4-fluoro-2-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

76066-5G 76066-25G 76066-1G 76066-500MG
77630-662EA 1441 USD
77630-662 77636-208 77639-980 77651-996
5-Ethoxy-4-fluoro-2-methylbenzaldehyde ≥97%
5-Ethoxy-4-fluoro-2-methylbenzaldehyde

Precaución: For Research Use Only

Formula: C₁₀H₁₁FO₂
Storage Temperature: Ambient
MDL Number: MFCD34766239
CAS Number: 2092867-42-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR