5-Bromo-2-fluoro-1-methoxy-3-(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

75676-1G 75676-5G 75676-500MG
77639-978EA 684 USD
77639-978 77660-320 77647-962
5-Bromo-2-fluoro-1-methoxy-3-(trifluoromethyl)benzene ≥97%
5-Bromo-2-fluoro-1-methoxy-3-(trifluoromethyl)benzene

Precaución: For Research Use Only

Formula: 273.0223 g/mol
MW: C₈H₅BrF₄O
Storage Temperature: Ambient
MDL Number: MFCD32694937
CAS Number: 2092799-62-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR