Tert-butyl 3-bromo-2,6-difluorobenzoate ≥95%

Supplier: AOBChem USA

Warning

75654-5G 75654-25G 75654-1G 75654-500MG
77630-660EA 1441 USD
77630-660 77636-206 77644-000 77651-994
Tert-butyl 3-bromo-2,6-difluorobenzoate ≥95%
Tert-butyl 3-bromo-2,6-difluorobenzoate

Precaución: For Research Use Only

Formula: C₁₁H₁₁BrF₂O₂
Storage Temperature: Ambient
MDL Number: MFCD35165157
CAS Number: 3002502-34-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR