4-Chloro-3-methoxy-2-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

75536-5G 75536-1G 75536-500MG
77660-318EA 2565 USD
77660-318 77643-998 77651-992
4-Chloro-3-methoxy-2-methylbenzaldehyde ≥97%
4-Chloro-3-methoxy-2-methylbenzaldehyde

Precaución: For Research Use Only

Formula: C₉H₉ClO₂
Storage Temperature: Ambient
MDL Number: MFCD34766055
CAS Number: 2384258-55-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR