5-Iodo-[1,1'-biphenyl]-2-ol ≥95%

Supplier: AOBChem USA

Warning

75414-5G 75414-25G 75414-1G 75414-500MG
77635-296EA 268 USD
77635-296 77656-874 77643-996 77647-956
5-Iodo-[1,1'-biphenyl]-2-ol ≥95%
5-Iodo-[1,1'-biphenyl]-2-ol

Precaución: For Research Use Only

Formula: C₁₂H₉IO
MW: 296.1 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11110537
CAS Number: 87441-19-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR