1-(3-Chloro-2-fluoro-5-methylphenyl)ethanol ≥95%

Supplier: AOBChem USA

Warning

75216-1G 75216-5G 75216-500MG
77639-972EA 551 USD
77639-972 77656-872 77647-954
1-(3-Chloro-2-fluoro-5-methylphenyl)ethanol ≥95%
1-(3-Chloro-2-fluoro-5-methylphenyl)ethanol

Precaución: For Research Use Only

Formula: 188.6265 g/mol
MW: C₉H₁₀ClFO
Storage Temperature: Ambient
MDL Number: MFCD28522711
CAS Number: 1780160-23-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR