1-Chloro-2-fluoro-3-iodo-5-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

75153-1G 75153-5G 75153-500MG
77635-294EA 1096 USD
77635-294 77656-870 77647-952
1-Chloro-2-fluoro-3-iodo-5-methylbenzene ≥95%
1-Chloro-2-fluoro-3-iodo-5-methylbenzene

Precaución: For Research Use Only

Formula: 270.47 g/mol
MW: C₇H₅ClFI
Storage Temperature: Ambient
MDL Number: MFCD28751666
CAS Number: 1805952-97-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR