1-(3-Chloro-2-fluoro-5-methylphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

75149-5G 75149-1G 75149-500MG
77630-656EA 1788 USD
77630-656 77643-992 77651-988
1-(3-Chloro-2-fluoro-5-methylphenyl)ethanone ≥95%
1-(3-Chloro-2-fluoro-5-methylphenyl)ethanone

Precaución: For Research Use Only

Formula: 186.611 g/mol
MW: C₉H₈ClFO
Storage Temperature: Ambient
MDL Number: MFCD28522255
CAS Number: 1783338-12-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR