(3-Chloro-2-fluoro-5-methylphenyl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

75144-5G 75144-25G
77635-292EA 136 USD
77635-292 77656-868
(3-Chloro-2-fluoro-5-methylphenyl)boronic acid ≥97%
(3-Chloro-2-fluoro-5-methylphenyl)boronic acid

Precaución: For Research Use Only

Formula: C₇H₇BClFO₂
MW: 188.392 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22571810
CAS Number: 352535-88-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR