2-(Benzyloxy)-3,5-dibromo-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

74849-5G 74849-1G 74849-25G 74849-500MG
77630-652EA 1441 USD
77630-652 77639-968 77656-864 77647-946
2-(Benzyloxy)-3,5-dibromo-1,1'-biphenyl ≥97%
2-(Benzyloxy)-3,5-dibromo-1,1'-biphenyl

Precaución: For Research Use Only

Formula: 418.122 g/mol
MW: C₁₉H₁₄Br₂O
Storage Temperature: Ambient
MDL Number: MFCD34765633
CAS Number: 2807444-86-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR