1,5-Dichloro-2,4-difluoro-3-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

74570-5G 74570-1G 74570-25G 74570-500MG
77630-650EA 1441 USD
77630-650 77639-966 77656-862 77651-986
1,5-Dichloro-2,4-difluoro-3-methylbenzene ≥95%
1,5-Dichloro-2,4-difluoro-3-methylbenzene

Precaución: For Research Use Only

Formula: C₇H₄Cl₂F₂
Storage Temperature: Ambient
CAS Number: 3037682-27-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR