1-Fluoro-4-isobutoxy-2-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

74110-5G 74110-1G 74110-500MG 74110-25G
77630-648EA 1441 USD
77630-648 77639-964 77653-074 77656-860
1-Fluoro-4-isobutoxy-2-methylbenzene ≥97%
1-Fluoro-4-isobutoxy-2-methylbenzene

Precaución: For Research Use Only

Formula: 182.235 g/mol
MW: C₁₁H₁₅FO
Storage Temperature: Ambient
MDL Number: MFCD24104896
CAS Number: 1501175-70-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR