4-Chloro-3-fluoro-5-iodobenzoic acid ≥95%

Supplier: AOBChem USA

Warning

73851-5G 73851-25G 73851-1G 73851-500MG
77630-646EA 1441 USD
77630-646 77636-202 77643-988 77651-984
4-Chloro-3-fluoro-5-iodobenzoic acid ≥95%
4-Chloro-3-fluoro-5-iodobenzoic acid

Precaución: For Research Use Only

Formula: C₇H₃ClFIO₂
Storage Temperature: Ambient
MDL Number: MFCD34765166
CAS Number: 2383665-70-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR