1-Bromo-2-(difluoromethoxy)-3,5-dimethylbenzene ≥95%

Supplier: AOBChem USA

Warning

73550-5G 73550-25G 73550-1G 73550-500MG
77630-644EA 1468 USD
77630-644 77636-200 77639-962 77647-944
1-Bromo-2-(difluoromethoxy)-3,5-dimethylbenzene ≥95%
1-Bromo-2-(difluoromethoxy)-3,5-dimethylbenzene

Precaución: For Research Use Only

Formula: C₉H₉BrF₂O
Storage Temperature: Ambient
MDL Number: MFCD31587955
CAS Number: 2138358-26-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR