2,2,2-Trifluoro-1-(5-isopropyl-2-methoxyphenyl)ethanone ≥97%

Supplier: AOBChem USA

Warning

73525-5G 73525-1G 73525-25G 73525-500MG
77630-642EA 1441 USD
77630-642 77639-960 77656-856 77647-942
2,2,2-Trifluoro-1-(5-isopropyl-2-methoxyphenyl)ethanone ≥97%
2,2,2-Trifluoro-1-(5-isopropyl-2-methoxyphenyl)ethanone

Precaución: For Research Use Only

Formula: 246.226 g/mol
MW: C₁₂H₁₃F₃O₂
Storage Temperature: Ambient
MDL Number: MFCD07380792
CAS Number: 1368497-73-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR