1-Bromo-3-chloro-2-(difluoromethoxy)-4-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

73227-5G 73227-1G 73227-25G 73227-500MG
77630-640EA 1441 USD
77630-640 77639-958 77660-308 77647-940
1-Bromo-3-chloro-2-(difluoromethoxy)-4-methylbenzene ≥97%
1-Bromo-3-chloro-2-(difluoromethoxy)-4-methylbenzene

Precaución: For Research Use Only

Formula: 271.486 g/mol
MW: C₈H₆BrClF₂O
Storage Temperature: Ambient
MDL Number: MFCD34764832
CAS Number: 2387098-01-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR