1-(3-Chloro-4-methoxyphenyl)ethanol ≥95%

Supplier: AOBChem USA

Warning

72874-5G 72874-25G 72874-1G 72874-500MG
77633-704EA 1058 USD
77633-704 77660-306 77643-984 77651-980
1-(3-Chloro-4-methoxyphenyl)ethanol ≥95%
1-(3-Chloro-4-methoxyphenyl)ethanol

Precaución: For Research Use Only

Formula: C₉H₁₁ClO₂
Storage Temperature: Ambient
MDL Number: MFCD12786788
CAS Number: 124829-13-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR