3'-Chloro-4'-methoxyacetophenone ≥97%

Supplier: AOBChem USA

Warning

72873-5G 72873-25G
77635-288EA 59 USD
77635-288 77656-848
3'-Chloro-4'-methoxyacetophenone ≥97%
3'-Chloro-4'-methoxyacetophenone

Precaución: For Research Use Only

Formula: C₉H₉ClO₂
Storage Temperature: Ambient
MDL Number: MFCD04973413
CAS Number: 37612-52-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR