1-(4-Chloro-3-fluoro-2-methylphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

72732-5G 72732-25G 72732-1G 72732-500MG
77630-636EA 1441 USD
77630-636 77656-846 77643-982 77647-938
1-(4-Chloro-3-fluoro-2-methylphenyl)ethanone ≥95%
1-(4-Chloro-3-fluoro-2-methylphenyl)ethanone

Precaución: For Research Use Only

Formula: 186.611 g/mol
MW: C₉H₈ClFO
Storage Temperature: Ambient
MDL Number: MFCD28555001
CAS Number: 1780212-07-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR