2,2,2-Trifluoro-1-(2-methoxynaphthalen-1-yl)ethanone ≥95%

Supplier: AOBChem USA

Warning

72639-5G 72639-1G 72639-500MG
77630-634EA 1537 USD
77630-634 77639-954 77651-978
2,2,2-Trifluoro-1-(2-methoxynaphthalen-1-yl)ethanone ≥95%
2,2,2-Trifluoro-1-(2-methoxynaphthalen-1-yl)ethanone

Precaución: For Research Use Only

Formula: C₁₃H₉F₃O₂
Storage Temperature: Ambient
MDL Number: MFCD02260833
CAS Number: 169295-61-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR