(4-Chloro-2-methylphenyl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

72589-5G 72589-25G 72589-1G 72589-500MG
77633-702EA 1180 USD
77633-702 77660-302 77643-980 77647-936
(4-Chloro-2-methylphenyl)(methyl)sulfane ≥95%
(4-Chloro-2-methylphenyl)(methyl)sulfane

Precaución: For Research Use Only

Formula: 172.675 g/mol
MW: C₈H₉ClS
Storage Temperature: Ambient
MDL Number: MFCD11617796
CAS Number: 144266-84-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR