1,2-Dichloro-4-iodo-3-(methoxymethoxy)benzene ≥97%

Supplier: AOBChem USA

Warning

72429-5G 72429-1G 72429-25G 72429-500MG
77630-632EA 1441 USD
77630-632 77639-952 77656-844 77651-972
1,2-Dichloro-4-iodo-3-(methoxymethoxy)benzene ≥97%
1,2-Dichloro-4-iodo-3-(methoxymethoxy)benzene

Precaución: For Research Use Only

Formula: C₈H₇Cl₂IO₂
Storage Temperature: Ambient
MDL Number: MFCD34764356
CAS Number: 2901107-04-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR