1-(Benzo[b]thiophen-7-yl)-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

72116-5G 72116-1G 72116-500MG
77660-300EA 1946 USD
77660-300 77643-978 77651-968
1-(Benzo[b]thiophen-7-yl)-2,2-difluoroethanone ≥95%
1-(Benzo[b]thiophen-7-yl)-2,2-difluoroethanone

Precaución: For Research Use Only

Formula: C₁₀H₆F₂OS
Storage Temperature: Ambient
MDL Number: MFCD26045308
CAS Number: 1565398-83-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR