2,2-Difluoro-1-(2-fluoro-[1,1'-biphenyl]-4-yl)ethanone ≥95%

Supplier: AOBChem USA

Warning

72082-1G 72082-5G 72082-500MG
77639-950EA 577 USD
77639-950 77656-842 77651-966
2,2-Difluoro-1-(2-fluoro-[1,1'-biphenyl]-4-yl)ethanone ≥95%
2,2-Difluoro-1-(2-fluoro-[1,1'-biphenyl]-4-yl)ethanone

Precaución: For Research Use Only

Formula: 250.216 g/mol
MW: C₁₄H₉F₃O
Storage Temperature: Ambient
MDL Number: MFCD28103858
CAS Number: 2173093-63-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR