2,2-Difluoro-1-(4-methoxy-3-(trifluoromethyl)phenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

72081-1G 72081-5G 72081-500MG
77639-948EA 577 USD
77639-948 77660-298 77651-964
2,2-Difluoro-1-(4-methoxy-3-(trifluoromethyl)phenyl)ethanone ≥95%
2,2-Difluoro-1-(4-methoxy-3-(trifluoromethyl)phenyl)ethanone

Precaución: For Research Use Only

Formula: C₁₀H₇F₅O₂
Storage Temperature: Ambient
MDL Number: MFCD28104499
CAS Number: 2173116-42-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR