2,2-Difluoro-1-(4-fluoro-3-methoxyphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

71991-5G 71991-1G 71991-500MG
77656-840EA 2001 USD
77656-840 77643-976 77647-934
2,2-Difluoro-1-(4-fluoro-3-methoxyphenyl)ethanone ≥95%
2,2-Difluoro-1-(4-fluoro-3-methoxyphenyl)ethanone

Precaución: For Research Use Only

Formula: 204.146 g/mol
MW: C₉H₇F₃O₂
Storage Temperature: Ambient
MDL Number: MFCD28103895
CAS Number: 1822671-04-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR