2,2-Difluoro-1-(2,3,5,6-tetrafluorophenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

71970-5G 71970-1G 71970-500MG
77660-292EA 1921 USD
77660-292 77643-974 77651-960
2,2-Difluoro-1-(2,3,5,6-tetrafluorophenyl)ethanone ≥95%
2,2-Difluoro-1-(2,3,5,6-tetrafluorophenyl)ethanone

Precaución: For Research Use Only

Formula: 228.091 g/mol
MW: C₈H₂F₆O
Storage Temperature: Ambient
MDL Number: MFCD34815570
CAS Number: 1852000-47-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR