1-(2-Chloro-3,6-difluorophenyl)-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

71949-1G 71949-5G 71949-500MG
77639-942EA 753 USD
77639-942 77656-838 77647-932
1-(2-Chloro-3,6-difluorophenyl)-2,2-difluoroethanone ≥95%
1-(2-Chloro-3,6-difluorophenyl)-2,2-difluoroethanone

Precaución: For Research Use Only

Formula: C₈H₃ClF₄O
Storage Temperature: Ambient
MDL Number: MFCD30628753
CAS Number: 2141900-32-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR