2,2-Difluoro-1-(2-methyl-5-(trifluoromethyl)phenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

71947-5G 71947-1G 71947-500MG 71947-25G
77630-630EA 1441 USD
77630-630 77639-940 77653-070 77660-288
2,2-Difluoro-1-(2-methyl-5-(trifluoromethyl)phenyl)ethanone ≥95%
2,2-Difluoro-1-(2-methyl-5-(trifluoromethyl)phenyl)ethanone

Precaución: For Research Use Only

Formula: C₁₀H₇F₅O
Storage Temperature: Ambient
MDL Number: MFCD34815563
CAS Number: 2164967-65-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR