2,2-Difluoro-1-(3-phenoxyphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

71938-5G 71938-1G 71938-500MG
77630-628EA 1770 USD
77630-628 77639-938 77653-068
2,2-Difluoro-1-(3-phenoxyphenyl)ethanone ≥95%
2,2-Difluoro-1-(3-phenoxyphenyl)ethanone

Precaución: For Research Use Only

Formula: 248.225 g/mol
MW: C₁₄H₁₀F₂O₂
Storage Temperature: Ambient
MDL Number: MFCD34815561
CAS Number: 2228347-29-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR