2,2-Difluoro-1-(5-fluoro-2-methylphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

71870-5G 71870-25G 71870-1G 71870-500MG
77633-700EA 1024 USD
77633-700 77660-286 77643-970 77651-956
2,2-Difluoro-1-(5-fluoro-2-methylphenyl)ethanone ≥95%
2,2-Difluoro-1-(5-fluoro-2-methylphenyl)ethanone

Precaución: For Research Use Only

Formula: 188.146 g/mol
MW: C₉H₇F₃O
Storage Temperature: Ambient
MDL Number: MFCD20921036
CAS Number: 1352205-82-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR