1-(2,5-Diethylphenyl)-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

71847-5G 71847-1G 71847-25G 71847-500MG
77633-698EA 1357 USD
77633-698 77639-936 77656-836 77651-954
1-(2,5-Diethylphenyl)-2,2-difluoroethanone ≥95%
1-(2,5-Diethylphenyl)-2,2-difluoroethanone

Precaución: For Research Use Only

Formula: 212.236 g/mol
MW: C₁₂H₁₄F₂O
Storage Temperature: Ambient
MDL Number: MFCD34815535
CAS Number: 1697530-54-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR