2,2-Difluoro-1-(2-methoxy-3-methylphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

71771-5G 71771-1G 71771-500MG
77660-284EA 1921 USD
77660-284 77643-968 77647-930
2,2-Difluoro-1-(2-methoxy-3-methylphenyl)ethanone ≥95%
2,2-Difluoro-1-(2-methoxy-3-methylphenyl)ethanone

Precaución: For Research Use Only

Formula: 200.182 g/mol
MW: C₁₀H₁₀F₂O₂
Storage Temperature: Ambient
MDL Number: MFCD34815516
CAS Number: 2229231-45-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR