1-(3-Chloro-5-methylphenyl)-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

71672-5G 71672-1G 71672-25G 71672-500MG
77630-626EA 1441 USD
77630-626 77639-934 77656-834 77651-952
1-(3-Chloro-5-methylphenyl)-2,2-difluoroethanone ≥95%
1-(3-Chloro-5-methylphenyl)-2,2-difluoroethanone

Precaución: For Research Use Only

Formula: C₉H₇ClF₂O
Storage Temperature: Ambient
MDL Number: MFCD34764178
CAS Number: 2354273-01-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR